Learn More
Enterprise Technology Solutions ProGuard Safety Goggles, w/Vent Caps, UV Protection, 12/BX, CL
SDP
Catalog No. 5000827997
Encompass_Preferred
Safety goggles help you maintain excellent visibility while protecting you from chemical splash. Indirect vent and cap vents allow air to flow through to prevent fogging. Soft PVC body is lightweight and comfortable for extended wear. Adjustable elastic headband provides a secure fit to prevent sliding off during use. Large design fits over most prescription eyewear. Safety goggles exceed ANSI z87.1-2003 high impact requirements.
Catalog No. 50-008-27997 Supplier Enterprise Technology Solutions Supplier No. PGD7321
Safety goggles help you maintain excellent visibility while protecting you from chemical splash. Indirect vent and cap vents allow air to flow through to prevent fogging. Soft PVC body is lightweight and comfortable for extended wear. Adjustable elastic headband provides a secure fit to prevent sliding off during use. Large design fits over most prescription eyewear. Safety goggles exceed ANSI z87.1-2003 high impact requirements.