Learn More
Medchemexpress LLC HY-K0221 1 mL Medchemexpress, Affinity Chromatography Column CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYK02211 ML10 PCS
Encompass_Preferred
Medchemexpress, HY-P7400 10ug Proheparin-binding EGF-like Growth Factor (CHO-expressed), Human CAS: Recombinant Human Proheparin-binding EGF-like Growth Factor (CHO-expressed) is shown to play a role in wound healing, cardiac hypertrophy, and heart development and function. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0205