Learn More
Cayman Chemical Prohexadione, Purity: ≥98%, CAS: 88805-35-0, 10 mg
SDP
Catalog No. 502947037
Encompass_Preferred
A plant growth regulator; inhibits the gibberellin plant hormone biosynthesis enzyme 3β-hydroxylase at 0.1, 1, and 10 mM; reduces the shoot length of peanut plants when applied at 0.5, 1.5, and 4.5 kg/hectare; inhibits JMJD2A demethylase at 1 mM; induces the differentiation of mouse neurospheres into neurons at 1, 1.5, and 2 mM
Catalog No. 50-294-7037 Supplier Cayman Chemical Supplier No. 3392210 MG
A plant growth regulator; inhibits the gibberellin plant hormone biosynthesis enzyme 3β-hydroxylase at 0.1, 1, and 10 mM; reduces the shoot length of peanut plants when applied at 0.5, 1.5, and 4.5 kg/hectare; inhibits JMJD2A demethylase at 1 mM; induces the differentiation of mouse neurospheres into neurons at 1, 1.5, and 2 mM