Learn More
CPC Scientific H-Arg-Arg-Glu-Ala-Glu-Asp-Leu-Gln-Val-Gly-Gln-Val-Glu-Leu-Gly-Gly-Gly-Pro-Gly-Ala-Gly-Ser-Leu-Gln-Pro-Leu-Ala-Leu-Glu-Gly-Ser-Leu-Gln-Lys-Arg-OH 1MG
SDP
Supplier:  CPC Scientific CPEP005B
Encompass
ONE-LETTER SEQUENCE: RREAEDLQVGQVELGGGPGAGSLQPLALEGSLQKRMOLECULAR FORMULA: C153H259N49O52MOLECULAR WEIGHT:3617.1STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [11097-48-6], RESEARCH AREA: DiabetesREFERENCES: B.H. Frank et al., Peptides, 7th Amer. Pept. Symp., 729 (1981)
Catalog No. 50-210-9156