Learn More
Drg International Inc Proinsulin Elisa
Supplier:  Drg International Inc EIA1560
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of proinsulin (intact) in serum and plasma. The measurement of Proinsulin in serum can provide valuable information for the diagnosis of insulinomas. Proinsulin levels have also been shown to be elevated in non-insulin dependent diabetics (NIDDM), newly diagnosed insulin dependent diabetics (IDDM) and other clinical situations. The DRG Proinsulin ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal antibody directed towards a unique antigenic site on a Proinsulin molecule.
Catalog No. 50-194-7537