Learn More
Mercodia Inc Proinsulin ELISA, Human, Serum Plasma, 96 Wells
SDP
Supplier:  Mercodia Inc 10111801
Encompass_Preferred
Mercodia Proinsulin ELISA provides a method for the quantitative determination of human proinsulin in serum, plasma, or cell culture media. Highly specific for proinsulin, with no cross-reactivity to c-peptide or insulin.
Catalog No. NC0027871