Learn More
IBL America Proinsulin (Intact) ELISA
Supplier:  IBL America IV2102E
Encompass_Preferred
Enzyme immunoassay (ELISA) for the quantitative measurement of intact proinsulin in heparin or EDTA plasma. For research use only, not for use in diagnostic procedures.The sample is incubated in the microtitre well togetherwith a buffer and, after a wash step, the biotin labelled antibody solution is added. After asecond incubation and wash step, HRP labelled streptavidin is added. A third incubation andwash is followed by the addition of substrate solution. Following colour development “stopreagent” is added and the colour intensity measured in a 96-well microplate reader.
Catalog No. NC1491393