Learn More
GSC Go Science Crazy Projectile Launcher
SCI_ED
Catalog No. S13995 Shop All GSC Products
Encompass
Encompass_Preferred
Catalog No. S13995 Supplier GSC Go Science Crazy Supplier No. PRLCH01
Alter the mass of projectiles and observe the effect on their trajectories. The GSC Projectile Launcher features a built-in clamp to fasten the device securely to tabletops.
  • Students can alter the mass of the ball and observe the effect on the ball's trajectory
  • The launcher features a spring loaded trigger and easy to use release mechanism that can be adjusted for tension
  • A protractor and plumb bob are fixed to the face of the apparatus for determining the angle of inclination
  • The built-in clamp is used to secure the apparatus to a tabletop
  • Four holes spaced 5cm apart are located at the top and bottom edge and are used for attaching a photogate (not included)
  • The kit also included three 25mm balls with a different mass, aluminum, lead and steel
Specifications
Product Type Demonstration
Includes Launcher, built-in clamp, and three 25mm balls; aluminum, lead, and steel
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov