Learn More
3B Scientific™ Projectile Launcher
SDPSCI_ED
Catalog No. S21116 Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
1
Catalog No. Quantity
S21116 1
1 options
Catalog No. S21116 Supplier 3B Scientific™ Supplier No. U10360
Add to Cart
Add to Cart
3B Scientific™ Projectile Launcher is for the quantitative investigation of projectile laws (vertical, horizontal and angled launch) and recording flight trajectories depending on launch angle and projectile range
  • Projectile is launched with almost no spin.
  • Determination of range and height from the projection angle
  • Determination of launch speed from the range and height
  • Determination of launch speed using a ballistic pendulum
  • Elastic and inelastic collisions
Specifications
Product Type Physics Equipment
Includes Launcher, Projectiles, Ramrod, Wing nut
Width (Metric) 15 cm
Height (Metric) 21 cm
Quantity 1
Material Metal, Plastic
Color White
Languages English