Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Projectile Launcher for Plastic Marbles
SCI_ED

Catalog No. S38982 Shop All GSC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S38982 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S38982 Supplier GSC Supplier No. PROLCH
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSC Projectile Launcher for Plastic Marbles can launch the plastic marbles a maximum of 8 meters

  • Launch tube has five speed settings that can lock into place to assure consistent launches at the same velocities
  • Protractor printed on the base to measure the angle of launch
  • Photogate (not included) can be attached near the mouth of the launcher to accurately record initial velocity

Specifications

Product Type Physical Science
Includes Launcher, Base, 2 marbles
Quantity 1
Length (Metric) 42.5 cm
Width (Metric) 32.5 cm
Weight (Metric) 2.5 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.