Learn More
Epigentek Inc Prok2 Polyclonal Antibody, HRP Conjulated (50 ul)
SDP
Supplier:  Epigentek Inc A67651050
Encompass_Preferred
HRP Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Mouse Prokineticin-2 protein (27-128AA). Tested Applications: ELISA. Reactivity: Mouse. Alternative Names: Prokineticin-2, PK2, Protein Bv8 homolog, Prok2, Bv8.
Catalog No. 50-124-9311