Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ ProKlenz 120 High Performance Alkaline Detergent

Catalog No. 17183983 Shop All STERIS Corporation Products
Change view
Click to view available options
Size:
5 gal.
Container Type:
Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Container Type
17-183-983 5 gal. Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-183-983 Supplier STERIS™ Supplier No. 143205WR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ProKlenz 120 Mild Alkaline Detergent is formulated to meet the unique cleaning challenges found in the pharmaceutical and other industries. ProKlenz 120 Detergent’s low foaming and free rinsing formula can be used in high-energy spray applications or manual cleaning. pH (1%): 11.5, typical.

ProKlenz™ 120 Alkaline Detergent is specially formulated to deliver effective cleaning with excellent substrate compatibility in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

How ProKlenz 120 Works:

ProKlenz 120 Alkaline Detergent is a phosphate-free, liquid, alkaline detergent for use in clean-in-place (CIP), clean-out-of-place (COP), and manual cleaning applications. It is formulated for enhanced materials compatibility.

pH (1%): 11.5, typical

Why ProKlenz 120?

  • Formulated alkaline detergent for effective removal of residues in both soft and hard water
  • Formulated for excellent materials compatibility
  • Reduces utility consumption and cleaning time with low foaming, free-rinsing formulation
  • Phosphate-free formulation with biodegradable surfactants
  • Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility.

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 11.5 (Diluted)
Scent Unscented
Sterility Non-sterile
Container Type Pail
Size 5 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.