Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS ProKlenz™ Booster

Catalog No. 19415172 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
5 gal.
55 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-415-172 5 gal.
19-415-173 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-415-172 Supplier STERIS Supplier No. 160905
Add to Cart
Add to Cart

Booster High Performance Detergent Additive has a mild pH is especially effective for the removal of tablet coatings in the pharmaceutical industry.

ProKlenz™ Booster High Performance Detergent Additive can resolve your most challenging process cleaning challenges by greatly enhancing the performance of other detergents with a state-of-the-art surfactant system and oxidizing chemistry.

This Detergent Additive is a proprietary blend of surfactants, an oxidizing agent and other performance-enhancing ingredients that provide multiple cleaning mechanisms to remove a wide range of process residues and is ideal for use in CIP, COP, and manual applications.

  • Enhances cleaning performance of other detergents or works individually with its superior blend of surfactants and oxidizing chemistry
  • Excellent product for tablet coating removal in pharmaceutical and dietary supplement manufacturing facilities
  • Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics (above 50°C) when used in clean-in-place (CIP), clean-out-of-place (COP), and manual applications
  • Phosphate-free, and manufactured with biodegradable ingredients to support harmonization of cleaning processes
  • Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
  • Low odor
  • Specific Gravity: 77°F (25°C) 1.040, typical
  • Solubility: Complete
  • Foam: Low at temperatures above 50°C

Specifications

Product Type Detergent Booster
pH 5.5 (undiluted)
Appearance Clear to Straw
Includes Pail
Volume (English) 5 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.