Learn More
STERIS ProKlenz™ Booster Sterile Detergent
Catalog No. 19415135 Shop All STERIS Corporation Products
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
19-415-135 Sterile
1 options
This item is not returnable. View return policy
Catalog No. 19-415-135 Supplier STERIS Supplier No. 1S1337WR
Add to Cart
Add to Cart
This item is not returnable. View return policy
Booster Sterile Detergent is a concentrated, neutral detergent for the removal of residues from hard surfaces in critical environments.
STERIS™ ProKlenz™ Booster Sterile Detergent works to remove residues from even the most critical of hard surfaces. This detergent is designed to remove organic and inorganic residues from product and nonproduct contact surfaces. Sterile filtered and filled in an ISO Class 5 aseptic environment, each package is double-bagged for easy entry into your controlled environments and aseptic core areas.
This product has an extensive documentation package to meet your validation objectives and is supported by the STERIS™ Technical Services team for individualized application and validation assistance.
  • Formulated with hydrogen peroxide and surfactants to utilize multiple cleaning mechanisms to clean a broad spectrum of soils
  • Moderate pH allows for safe use in manual cleaning and compatible with many substrates
  • Double-bagged for easy introduction into aseptic areas
  • Being 0.2 micron filtered, aseptically filled and packaged minimizes potential for particles being introduced during use
  • Each lot documented by a Certificate of Analysis including sterility results
  • Designed to be diluted to 1% v/v using sterile water for injection (WFI) or other high quality grade water
Specifications
Product Type Detergent Booster
Sterility Sterile
Quantity 24 Pk.
Volume (English) 2.6 oz.
These products are not registered as disinfectants by the US EPA.