Learn More
STERIS ProKlenz NpH Neutral Detergent
Catalog No. 17010354 Shop All STERIS Corporation Products
Change view
Click to view available options
Size:
5 gal.
55 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-354 5 gal.
17-010-353 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-010-354 Supplier STERIS Supplier No. 143005WR
Add to Cart
Add to Cart
ProKlenz NpH High Performance Neutral Process and Research Cleaner is formulated with surfactants, solvents, and other essential ingredients to utilize multiple cleaning mechanisms while enhancing substrate compatibility.
  • ProKlenz NpH High Performance Neutral Detergent is specially formulated to deliver effective cleaning with excellent substrate compatibility in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.
  • ProKlenz NpH Neutral Detergent is a proprietary blend of surfactants, solvents and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues and is ideal for use in CIP, COP, and manual applications.
  • Neutral formulation delivers excellent cleaning characteristics and substrate compatibility with enhanced operator safety
  • Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics when used in clean-in-place (CIP), clean-out-of-place (COP), and manual applications
  • Phosphate free, and manufactured with biodegradable ingredients to support harmonization of cleaning processes
  • Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
Specifications
Product Type Detergent
Color-coded No
Form Clear Liquid
Scent Unscented
Sterility Non-sterile
Size 5 gal.