Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ ProKlenz ONE High Performance Alkaline Cleaner

Catalog No. 17103049 Shop All STERIS Corporation Products
Change view
Click to view available options
Size:
5 gal.
55 gal.
Container Type:
Drum
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Container Type
17-103-049 55 gal. Drum
17-103-048 5 gal. Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-103-049 Supplier STERIS™ Supplier No. 142101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

Alkaline, Cleaner,

ProKlenz ONE: Low-foaming, NaOH-based, alkaline cleaner made with biodegradable ingredients for pharmaceutical, biotechnology, and cosmetic industries. Effective against biofilm, disinfection, and virucidal activity. Globally available and reliable. pH - (1%) 12.5, typical.

ProKlenz ONE High Performance Alkaline Cleaner is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement and other industries requiring an effective validatable cleaner.

HOW PROKLENZ ONE WORKS

ProKlenz ONE Cleaner is a proprietary blend of sodium hydroxide, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants, and is ideal for use in clean-in-place (CIP) and clean-out-of-place (COP).

  • Biofilm Removal: ProKlenz ONE Alkaline Cleaner is effective for biofilm removal when used at a 5% concentration of product per gallon of water on a pre-cleaned, non-porous, nonfood contact surface at 60°C for a contact time of 10 minutes.
  • Disinfection: ProKlenz ONE Alkaline Cleaner is effective against Staphylococcus aureus, Salmonella enterica, and Pseudomonas aeruginosa when used at a 1% concentration of product per gallon of water in 5% blood serum and 250 ppm of hard water at 60°C for a contact time of 5 minutes.
  • Virucidal Activity: ProKlenz ONE Alkaline Cleaner is effective against Poliovirus (type 1) when used at a 1% concentration of product per gallon of water in 5% blood serum and 250 ppm of hard water at 60°C for a contact time of 10 minutes.

WHY PROKLENZ ONE?

  • Productivity - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP) and clean-out-of-place (COP).
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Efficacy - Delivers microbial control in critical manufacturing processes with disinfectant, virucidal and biofilm removal capabilities.
  • Globally Availability - Manufactured with biodegradable ingredients to support harmonization of cleaning processes.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.

Order Info

Alkaline, Cleaner,

Certifications

Alkaline, Cleaner,

Compliance

Alkaline, Cleaner,

Warranty and Services

Alkaline, Cleaner,

Alkaline, Cleaner,

Specifications

Product Type Alkaline Cleaner
Color-coded No
Form Clear Liquid
pH 12.5 (Diluted)
Scent Unscented
Sterility Non-sterile
Container Type Drum
Size 55 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.