Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS ProKlenz™ Sterile Detergent

Catalog No. 19415136 Shop All STERIS Corporation Products
Change view
Click to view available options
Product Type:
Booster Detergent
NpH Detergent
Includes:
128 oz. (1 Gallon)
2.6 oz.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
19-415-136 Booster Detergent 128 oz. (1 Gallon)
19-415-133 NpH Detergent 2.6 oz.
19-415-134 NpH Detergent 128 oz. (1 Gallon)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-415-136 Supplier STERIS Supplier No. 1S1308WR
Add to Cart
Add to Cart

A concentrated detergent for use in controlled environments

  • Designed to remove organic and inorganic residues from product and non-product contact surfaces
  • Formulated with multiple surfactants to clean a broad spectrum of soils
  • Neutral pH allows for use in manual cleaning and compatibility with many substrates
  • Filtered and filled in an ISO Class 5 aseptic environment to minimize the potential of particles being introduced during use
  • Packages are double bagged for easy entry into controlled environments and aseptic cores
  • Includes an extensive documentation package to meet validation objectives
  • Each lot documented by a Certificate of Analysis that includes sterility results
  • Supported by the STERIS Technical Services team for individualized application and validation assistance

Specifications

Product Type Booster Detergent
pH Neutral
Sterility Sterile
Description Formulated with Hydrogen Peroxide
Filtered Filtered
For Use With (Application) Removing Organic/Inorganic Residues From Product or Non-Product Contact Surfaces
Includes 128 oz. (1 Gallon)
Quantity 4/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.