Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS ProKlenz™ TWO High Performance Acid Detergent

Catalog No. 19415161 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-415-161 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-415-161 Supplier STERIS Supplier No. 142505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This High Performance Acid Detergent is an organic acid blend of biodegradable ingredients designed for routine cleaning, rouge removal, and passivation.

ProKlenz™ TWO High Performance Acid Detergent is a specially formulated phosphate-free for routine cleaning, derouging and passivation in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

How ProKlenz™ TWO Works

ProKlenz™ TWO Acid Detergent is a proprietary blend of organic acids, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues, inorganic soils and rouge, and is ideal for use in CIP, COP, manual and passivation applications.

Why ProKlenz™ TWO?

  • Productivity
    • Effective for routine removal of inorganic soils and rouge as well as for passivation in clean-in-place (CIP), clean-out-of-place (COP), and manual applications.
  • Easy to Validate
    • Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Cost Savings
    • Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
  • Globally Availability
    • Phosphate-free, and manufactured with biodegradable ingredients to support harmonization of cleaning processes.
  • Reliability
    • Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.
  • Solubility
    • Complete
  • Foam
    • Low
  • Rinsing
    • Excellent
  • Phosphates
    • None; 0% P

Specifications

Product Type Acid Cleaner/Disinfectant
pH 2.4
Sterility Non-sterile
Composition Liquid
Container Type Pail
For Use With (Application) Routine Cleaning, Rouge Removal, Passivation
Volume (English) 5 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.