Learn More
Aviva Systems Biology Corp PROKR2 Antibody - C-terminal region (ARP64050_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64050_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PROKR2. It was validated on Western Blot. . Alternate names: GPR73L1, GPR73b, GPRg2, KAL3, PKR2, dJ680N4.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-156