Learn More
Eurofins Discoverx Prolactin, 10 µg. (92-1144)
Supplier:  Eurofins Discoverx 921144
Encompass
Prolactin is a pituitary hormone that promotes lactation, commonly referred to as the "milk hormone". Also has a wide range of regulatory functions on the reproductive and immune system, such as growth, differentiation, anti-apoptotic factors, and influences on hematopoiesis, angiogenesis, and blood clotting. Acts as an agonist for PRLR. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9676