Learn More
Enzo Life Sciences Prolactin ELISA Kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1610001
Encompass_Preferred
The Prolactin ELISA kit is a colorimetric immunoassay kit with results in 3 hours. Absorbance is read at 450 nm. Measure up to 38 samples in duplicate per 96 well  kit. Alternative name: Luteotropic hormone, Luteotropin. Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-201-3852