Learn More
Cusabio Technology LLC Prolactin/Luteotropic Hormone
Catalog No. 5000051936
Encompass_Preferred
This kit detects PRL/LTH in canine serum plasma and tissue homogenates using a competitive ELISA method offering a quantitative measurement range of 7 5 IU/mL to 3000 IU/mL with a sensitivity threshold of 7 5 IU/mL The assay requires 50-100 L of sample per test and can be completed within 1-5 hours utilizing absorbance detection at 450 nm to ensure reliable results for reproductive and endocrine function studies in veterinary research Its standardized protocol enables precise monitoring of prolactin/luteotropic hormone levels supporting diagnostic and experimental applications in canine biological samples
Catalog No. 50-000-51936 Supplier Cusabio Technology LLC Supplier No. CSBE14930C
This kit detects PRL/LTH in canine serum plasma and tissue homogenates using a competitive ELISA method offering a quantitative measurement range of 7 5 IU/mL to 3000 IU/mL with a sensitivity threshold of 7 5 IU/mL The assay requires 50-100 L of sample per test and can be completed within 1-5 hours utilizing absorbance detection at 450 nm to ensure reliable results for reproductive and endocrine function studies in veterinary research Its standardized protocol enables precise monitoring of prolactin/luteotropic hormone levels supporting diagnostic and experimental applications in canine biological samples