Learn More
ACROBiosystems Recombinant Protein;Human Prolactin R / PRLR Protein, Fc Tag ;HEK293;1MG;PRP,Prolactin R,PRLR,Prolactin receptor,PRL-R
Catalog No. 501658186
Encompass_Preferred
Recombinant Protein;Human Prolactin R / PRLR Protein, Fc Tag ;1MG;Human Prolactin R / PRLR Protein, Fc Tag (PRP-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Asp 234 (Accession # P16471-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.0 kDa. The protein migrates as 64 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Prolactin, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PRN-H5257) at 1 μg/mL (100 μL/well) can bind Human Prolactin R / PRLR Protein, Fc Tag (Cat. No. PRP-H5251) with a linear range of 2-39 ng/mL (QC tested).;Immobilized Human GH, Tag Free (Cat. No. GH1-H4117) at 2 μg/mL (100 μL/well) can bind Human Prolactin R / PRLR Protein, Fc Tag (Cat. No. PRP-H5251) with a linear range of 0.4-6 ng/mL (Routinely tested).
Catalog No. 50-165-8186 Supplier ACROBiosystems Supplier No. PRPH52511MG
Recombinant Protein;Human Prolactin R / PRLR Protein, Fc Tag ;1MG;Human Prolactin R / PRLR Protein, Fc Tag (PRP-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Asp 234 (Accession # P16471-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.0 kDa. The protein migrates as 64 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Prolactin, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PRN-H5257) at 1 μg/mL (100 μL/well) can bind Human Prolactin R / PRLR Protein, Fc Tag (Cat. No. PRP-H5251) with a linear range of 2-39 ng/mL (QC tested).;Immobilized Human GH, Tag Free (Cat. No. GH1-H4117) at 2 μg/mL (100 μL/well) can bind Human Prolactin R / PRLR Protein, Fc Tag (Cat. No. PRP-H5251) with a linear range of 0.4-6 ng/mL (Routinely tested).