Learn More
ACROBiosystems Recombinant Protein;Human Prolactin R / PRLR Protein, His Tag ;HEK293;1MG;PRR,Prolactin R,PRLR,Prolactin receptor,PRL-R
Supplier:  ACROBiosystems PRRH52HA1MG
Encompass_Preferred
Recombinant Protein;Human Prolactin R / PRLR Protein, His Tag ;1MG;Human Prolactin R / PRLR Protein, His Tag (PRR-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Asp 234 (Accession # P16471-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.3 kDa. The protein migrates as 35-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Prolactin, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PRN-H5257) at 5 μg/mL (100 μL/well) can bind Human Prolactin R / PRLR Protein, His Tag (Cat. No. PRR-H52Ha) with a linear range of 20-156 ng/mL (QC tested).;Immobilized Human GH, Tag Free (Cat. No. GH1-H4117) at 5 μg/mL (100 μL/well) can bind Human Prolactin R / PRLR Protein, His Tag (Cat. No. PRR-H52Ha) with a linear range of 0.313-5 μg/mL (Routinely tested).
Catalog No. 50-165-8137