Learn More
Pro-Lab Diagnostics™ Prolex™ - Blue Staph Latex Kits
SDP
Catalog No. 22286169 Shop All Pro-Lab Diagnostics Products
Change view
Click to view available options
Quantity:
100 Tests
300 Tests
48 Cards
Type:
Agglutination Card
Blue Staph Latex Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
22-286-169 48 Cards Agglutination Card
22-286-203 100 Tests Blue Staph Latex Kit
22-286-171 300 Tests Blue Staph Latex Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-286-169 Supplier Pro-Lab Diagnostics™ Supplier No. PL.09248
Add to Cart
Add to Cart
A rapid, sensitive method for distinguishing Staphylococcus aureus that possess coagulase (clumping factor) and/or protein A from other species of staphylococci
  • Rapid clear results within 20 seconds
  • Uses blue latex particles against a white background
Specifications
Detectable Analytes S. aureus Coagulase, Protein A
Type Agglutination Card
Quantity 48 Cards