Learn More
Cayman Chemical PROLI NONOate, Purity: ≥98%, CAS: 178948-42-0, 100 mg
SDP
Catalog No. 502896569
Encompass_Preferred
PROLI NONOate is a NO donor. It spontaneously dissociates in a pH-dependent, first-order process with a half-life of 1.8 seconds at 37°C (pH 7.4) to liberate 2 moles of NO per mole of parent compound.{3266}
Catalog No. 50-289-6569 Supplier Cayman Chemical Supplier No. 82145100 MG
PROLI NONOate is a NO donor. It spontaneously dissociates in a pH-dependent, first-order process with a half-life of 1.8 seconds at 37°C (pH 7.4) to liberate 2 moles of NO per mole of parent compound.{3266}