Learn More
Medchemexpress LLC Proline-2,3,3,4,4,5,5-d7 | 65807-21-8 | 98.0% | 122.17 | 250 MG
SDP
Catalog No. 5000366379
Encompass_Preferred
DL-Proline-d7 is a deuterium labeled biochemical reagent suitable for life science research. It is used as a biological material or organic compound, and is not for patient use. Deuteration, the incorporation of stable heavy isotopes, has potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-66379 Supplier Medchemexpress LLC Supplier No. 250MGHYW012908S
DL-Proline-d7 is a deuterium labeled biochemical reagent suitable for life science research. It is used as a biological material or organic compound, and is not for patient use. Deuteration, the incorporation of stable heavy isotopes, has potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.