Learn More
Pipette Supplies, Inc. PROLINE/EPET PIPETTE BATTERY
SDP
Catalog No. 501317648
Encompass
Pipette Supplies brand rechargeable NiMH battery for the Sartorius Proline Electronic and ePET, Eppendorf Response Model 4850, and Hamilton SofTouch single and multichannel pipettes.Maximum charging time for an empty battery is 12 hours. Battery must be charged prior to use. RoHS compliant.Don’t overpay! Identical to the OEM, except in a black wrap.Replacement battery for EP 022463004, HA 9168-01, and SA 711002.01.Made in China
Catalog No. 50-131-7648 Supplier Pipette Supplies, Inc. Supplier No. PS 711002
Pipette Supplies brand rechargeable NiMH battery for the Sartorius Proline Electronic and ePET, Eppendorf Response Model 4850, and Hamilton SofTouch single and multichannel pipettes.Maximum charging time for an empty battery is 12 hours. Battery must be charged prior to use. RoHS compliant.Don’t overpay! Identical to the OEM, except in a black wrap.Replacement battery for EP 022463004, HA 9168-01, and SA 711002.01.Made in China