Learn More
Eurofins Discoverx ProLink Cloning Vector Bundle, 4 vials, 10 µg. (93-0491)
Catalog No. 502000646
Encompass
The ProLink Cloning Vector Bundle is a bundle of four vectors for use in expressing a protein of interest fused to variations of the ProLink (PK) tag at the C-terminus. The PK tag is a version of ED used for EFC assays (in combination with EA). The ProLink 2 tag has a higher affinity for EA than ProLink 1, making it ideal for use with weaker protein:protein interactions. The ARMS (Arrestin Recruitment Modifying Sequence) PK tags contain GRK phosphorylation sites to improve ligand-dependent arrestin recruitment to GPCRs and improve assay performance. Provided as four vials, 1 vial each vector, of 10 μg vector in 20 μL liquid buffer. For detailed product information, User Manual and datasheet, visit www.discoverx.com/products/reagent/vector/cloning-vector-bundle-93-0491.
Catalog No. 50-200-0646 Supplier Eurofins Discoverx Supplier No. 930491
The ProLink Cloning Vector Bundle is a bundle of four vectors for use in expressing a protein of interest fused to variations of the ProLink (PK) tag at the C-terminus. The PK tag is a version of ED used for EFC assays (in combination with EA). The ProLink 2 tag has a higher affinity for EA than ProLink 1, making it ideal for use with weaker protein:protein interactions. The ARMS (Arrestin Recruitment Modifying Sequence) PK tags contain GRK phosphorylation sites to improve ligand-dependent arrestin recruitment to GPCRs and improve assay performance. Provided as four vials, 1 vial each vector, of 10 μg vector in 20 μL liquid buffer. For detailed product information, User Manual and datasheet, visit www.discoverx.com/products/reagent/vector/cloning-vector-bundle-93-0491.