Learn More
Medchemexpress LLC Prolyl endopeptidase inhibitor 1 (Boc-Pro-prolinal) | 86925-97-5 | 98.8% | 296.36 | 1 ML
SDP
Catalog No. 5000437699
Encompass_Preferred
Prolyl Endopeptidase Inhibitor 1 (Boc-Pro-prolinal) is a potent prolyl endopeptidase (PEP; PE) inhibitor with a Ki value of 15 nM. It also exhibits an anti-amnesic effect. It strongly inhibits mammalian prolyl endopeptidase with a Ki value in the nM range, while its Ki value for the microbial enzyme (Flavobacterium) is 3.2 μM.
  • Potent prolyl endopeptidase (PEP; PE) inhibitor with a Ki value of 15 nM.
  • Exhibits anti-amnesic effect.
  • Strongly inhibits mammalian prolyl endopeptidase with a Ki value in the nM range.
Catalog No. 50-004-37699 Supplier Medchemexpress LLC Supplier No. HY11395110MM/1ML
Prolyl Endopeptidase Inhibitor 1 (Boc-Pro-prolinal) is a potent prolyl endopeptidase (PEP; PE) inhibitor with a Ki value of 15 nM. It also exhibits an anti-amnesic effect. It strongly inhibits mammalian prolyl endopeptidase with a Ki value in the nM range, while its Ki value for the microbial enzyme (Flavobacterium) is 3.2 μM.
  • Potent prolyl endopeptidase (PEP; PE) inhibitor with a Ki value of 15 nM.
  • Exhibits anti-amnesic effect.
  • Strongly inhibits mammalian prolyl endopeptidase with a Ki value in the nM range.