Learn More
Medchemexpress LLC Prolyl Endopeptidase Inhibitor 1 (Boc-Pro-prolinal) | 86925-97-5 | 98.8% | 296.36 | 10 MG
SDP
Catalog No. 5000435790
Encompass_Preferred
Prolyl Endopeptidase Inhibitor 1 (Boc-Pro-prolinal) is a potent prolyl endopeptidase (PEP; PE) inhibitor with a Ki value of 15 nM, and it has an anti-amnesic effect. It strongly inhibits mammalian prolyl endopeptidase in the nM range, and the microbial enzyme with a Ki value of 3.2 μM for Flavobacterium.
  • Potent prolyl endopeptidase (PEP; PE) inhibitor.
  • Ki value of 15 nM for prolyl endopeptidase.
  • Has anti-amnesic effect.
  • Inhibits mammalian prolyl endopeptidase with nM order Ki.
  • Inhibits microbial enzyme (Flavobacterium) with a Ki value of 3.2 μM.
  • Appearance: Solid, white to off-white.
Catalog No. 50-004-35790 Supplier Medchemexpress LLC Supplier No. HY11395110MG
Prolyl Endopeptidase Inhibitor 1 (Boc-Pro-prolinal) is a potent prolyl endopeptidase (PEP; PE) inhibitor with a Ki value of 15 nM, and it has an anti-amnesic effect. It strongly inhibits mammalian prolyl endopeptidase in the nM range, and the microbial enzyme with a Ki value of 3.2 μM for Flavobacterium.
  • Potent prolyl endopeptidase (PEP; PE) inhibitor.
  • Ki value of 15 nM for prolyl endopeptidase.
  • Has anti-amnesic effect.
  • Inhibits mammalian prolyl endopeptidase with nM order Ki.
  • Inhibits microbial enzyme (Flavobacterium) with a Ki value of 3.2 μM.
  • Appearance: Solid, white to off-white.