Learn More
Aviva Systems Biology Corp PROM1 Antibody - C-terminal region (ARP61145_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61145_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PROM1. It was validated on Western Blot. . Alternate names: AC133, CD133, CORD12, MCDR2, MSTP061, PROML1, RP41, STGD4ApplicationWB. Species reactivity: Human
Catalog No. 50-287-270