Learn More
Scott Safety™ Promask 25 Facepiece
Catalog No. 19222928
Change view
Click to view available options
Size:
Small
Catalog No. Size
19-222-928 Small
1 options
This item is not returnable. View return policy
Catalog No. 19-222-928 Supplier Scott Safety™ Supplier No. 013027
This item is not returnable. View return policy
Promask 25 full facepiece respirators are ergonomically designed to be lightweight and well-balanced, suitable for use in any industrial or manufacturing environment.
  • Halo-butyl elastomer material offers unmatched resistance to chemicals and ozone aging
  • Built-in speech diaphragm improves speech intelligibility for overall clearer communication
  • Improves worker effectiveness and safety
  • Wide, polycarbonate lens offers increased viewing area
  • Cartridge placement is out of user's line of sight
  • Wider sealing edge on facepiece and adjustable 5-point rubber head harness, ensures secure fit on any face
  • Sweat port in chin pocket provides added comfort during extended wear
Specifications
Material Thermoplastic Elastomer
Product Type Full Facepiece Respirator
Size Small
Facepiece Material Thermoplastic Elastomer
Includes Speaking Diaphragm
Lens Type Wide
Material (Lens) Polycarbonate
Model 013027
Suspension Type 5-point