Learn More
Med Vet International Promate™ CL Cordless Dental Polishing Unit, J1632, Each
Catalog No. 5000585690
Encompass_Preferred
Promate™ CL Cordless Dental Polishing Unit. Designed to provide the clinician with an ergonomic handpiece that delivers independence from heavy cord-drag, dual-mode speed control, and operator comfort during prophylaxis procedures. • 20% lighter than industry-leading, low-speed handpieces • Dual-mode speed control with 6-speed settings • Ergonomic, quick-touch button and Bluetooth-powered foot pedal • Use all your favorite disposable prophy angles • Battery life lasts through a whole day's worth of procedures • Outer sheath designed to be steam autoclaved
Catalog No. 50-005-85690 Supplier Med Vet International Supplier No. JOR1632
Promate™ CL Cordless Dental Polishing Unit. Designed to provide the clinician with an ergonomic handpiece that delivers independence from heavy cord-drag, dual-mode speed control, and operator comfort during prophylaxis procedures. • 20% lighter than industry-leading, low-speed handpieces • Dual-mode speed control with 6-speed settings • Ergonomic, quick-touch button and Bluetooth-powered foot pedal • Use all your favorite disposable prophy angles • Battery life lasts through a whole day's worth of procedures • Outer sheath designed to be steam autoclaved