Learn More
Impact Products ProMax (Microporous) Coverall Zipper Front Long Sleeve Open Wrists and Ankles White, 5XL
Catalog No. NC1562506
Encompass_Preferred
A microporous fabric that is suitable for many commercial applications. Applications are: asbestos abatement, general maintenance, food processing with chemicals, painting and pesticide spraying.
This item is not returnable. View return policy
Catalog No. NC1562506 Supplier Impact Products Supplier No. M10125X
A microporous fabric that is suitable for many commercial applications. Applications are: asbestos abatement, general maintenance, food processing with chemicals, painting and pesticide spraying.