Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Promazine hydrochloride | 53-60-1 | 99.9% | 1 ML
SDP

Catalog No. 5000412942
Encompass_Preferred

Promazine hydrochloride is an antipsychotic and a dopamine receptor D2 antagonist that inhibits dopaminergic neurotransmission. It is supplied as a solid, white to off-white powder.

  • Antipsychotic and dopamine receptor D2 antagonist
  • Inhibits dopaminergic neurotransmission
  • Shows KDs of 25, 190, and 8400 nM at human norepinephrine, serotonin, and dopamine transporters
  • Molecular weight: 320.88
  • Chemical formula: C17H21ClN2S
  • Soluble in water and DMSO at 100 mg/mL

Catalog No. 50-004-12942 Supplier Medchemexpress LLC Supplier No. HYB122510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Promazine hydrochloride is an antipsychotic and a dopamine receptor D2 antagonist that inhibits dopaminergic neurotransmission. It is supplied as a solid, white to off-white powder.

  • Antipsychotic and dopamine receptor D2 antagonist
  • Inhibits dopaminergic neurotransmission
  • Shows KDs of 25, 190, and 8400 nM at human norepinephrine, serotonin, and dopamine transporters
  • Molecular weight: 320.88
  • Chemical formula: C17H21ClN2S
  • Soluble in water and DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.