Learn More
Cayman Chemical PromestrIen 10mg
SDP
Catalog No. 502948710
Encompass_Preferred
A synthetic estrogen; reduces vaginal atrophy and decreases in endometrial and myometrial wall thickness in ovariectomized rats when applied as a topical gel
Catalog No. 50-294-8710 Supplier Cayman Chemical Supplier No. 3342610 MG
A synthetic estrogen; reduces vaginal atrophy and decreases in endometrial and myometrial wall thickness in ovariectomized rats when applied as a topical gel