Learn More
Cayman Chemical PromestrIen 50mg
SDP
Catalog No. 502949730
Encompass_Preferred
A synthetic estrogen; reduces vaginal atrophy and decreases in endometrial and myometrial wall thickness in ovariectomized rats when applied as a topical gel
Catalog No. 50-294-9730 Supplier Cayman Chemical Supplier No. 3342650 MG
A synthetic estrogen; reduces vaginal atrophy and decreases in endometrial and myometrial wall thickness in ovariectomized rats when applied as a topical gel