Learn More
Medchemexpress LLC Promestriene | 39219-28-8 | MFCD00867247 | 99.9% | 328.49 | C22H32O2 | 10 MG
SDP
Catalog No. 5000242222
Encompass_Preferred
Promestriene is a synthetic diethyl ether derivative of estradiol supplied as a solid research reagent. It functions as a locally active estrogen and is used in in vitro and analytical studies of estrogen receptor activity and vaginal atrophy. The material is accompanied by quality documentation for research use only.
  • High purity (~99.9%) analytical standard.
  • Molecular weight 328.49 g/mol and formula C22H32O2 for accurate calculations.
  • Available in small solid quantities, including 10 mg, suitable for lab-scale experiments.
  • Supplied with a datasheet and batch information for quality control.
  • Intended for research and analytical use; not for clinical or human use.
Catalog No. 50-002-42222 Supplier Medchemexpress LLC Supplier No. HY10829310MG
Promestriene is a synthetic diethyl ether derivative of estradiol supplied as a solid research reagent. It functions as a locally active estrogen and is used in in vitro and analytical studies of estrogen receptor activity and vaginal atrophy. The material is accompanied by quality documentation for research use only.
  • High purity (~99.9%) analytical standard.
  • Molecular weight 328.49 g/mol and formula C22H32O2 for accurate calculations.
  • Available in small solid quantities, including 10 mg, suitable for lab-scale experiments.
  • Supplied with a datasheet and batch information for quality control.
  • Intended for research and analytical use; not for clinical or human use.