Learn More
Apexbio Technology LLC Promethazine HCl 58-33-3 1g
SDP
Catalog No. 5000090842
Encompass_Preferred
Promethazine HCl (CAS 58-33-3) is a phenothiazine-derived small molecule that functions primarily as an antagonist at histamine H1 receptors and muscarinic acetylcholine receptors It also demonstrates affinity for serotonergic 5-HT2A and 5-HT2C receptors Through these interactions Promethazine HCl modulates histaminergic and cholinergic signaling resulting in antihistaminic sedative anticholinergic and antiemetic effects This compound is widely employed in biomedical research to investigate mechanisms underlying allergic responses motion sickness emesis and cholinergic neurotransmission
Catalog No. 50-000-90842 Supplier Apexbio Technology LLC Supplier No. B47841000
Add to Cart
Add to Cart
Promethazine HCl (CAS 58-33-3) is a phenothiazine-derived small molecule that functions primarily as an antagonist at histamine H1 receptors and muscarinic acetylcholine receptors It also demonstrates affinity for serotonergic 5-HT2A and 5-HT2C receptors Through these interactions Promethazine HCl modulates histaminergic and cholinergic signaling resulting in antihistaminic sedative anticholinergic and antiemetic effects This compound is widely employed in biomedical research to investigate mechanisms underlying allergic responses motion sickness emesis and cholinergic neurotransmission