Learn More
Medchemexpress LLC Promethazine sulfoxide | 7640-51-9 | MFCD08063694 | 10mg
SDP
Catalog No. 5000269463
Encompass_Preferred
Promethazine sulfoxide is a metabolite of the histamine H1 receptor antagonist Promethazine (HY-B1296) It is formed from promethazine by the cytochrome P450 (CYP) isoform CYP2D6
Catalog No. 50-002-69463 Supplier Medchemexpress LLC Supplier No. HYZ910110MG
Promethazine sulfoxide is a metabolite of the histamine H1 receptor antagonist Promethazine (HY-B1296) It is formed from promethazine by the cytochrome P450 (CYP) isoform CYP2D6