Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nanotemper Technologies Inc PROMETHEUS NT.48 NANODSF GRADE

Catalog No. NC1187355
Encompass

  • Product type: Standard capillaries
  • Grade: nanoDSF Grade
  • Compatibility: Prometheus NT.48, Prometheus Panta
  • Quantity: 400 capillaries per pack (2 x 200)
  • Application: Thermal and chemical stability measurements
  • Detection method: Intrinsic tryptophan and tyrosine fluorescence
  • Concentration range: Few µg/mL up to 250 mg/mL
  • Label-free: true
  • Sample volume: ~10 µL
  • Material purity: High glass purity
  • Reproducibility: Exceptional
  • Signal detection: Robust
  • Maintenance: Low
  • Buffer compatibility: Independent of buffer composition
  • Detergent compatibility: Compatible with detergents

Catalog No. NC1187355 Supplier Nanotemper Technologies Inc Supplier No. PRC002
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Product type: Standard capillaries
  • Grade: nanoDSF Grade
  • Compatibility: Prometheus NT.48, Prometheus Panta
  • Quantity: 400 capillaries per pack (2 x 200)
  • Application: Thermal and chemical stability measurements
  • Detection method: Intrinsic tryptophan and tyrosine fluorescence
  • Concentration range: Few µg/mL up to 250 mg/mL
  • Label-free: true
  • Sample volume: ~10 µL
  • Material purity: High glass purity
  • Reproducibility: Exceptional
  • Signal detection: Robust
  • Maintenance: Low
  • Buffer compatibility: Independent of buffer composition
  • Detergent compatibility: Compatible with detergents

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.