Learn More
Medchemexpress LLC Prometon-d14 | 1219803-45-8 | 239.38 g·mol⁻¹ | C10H5D14N5O | 1 MG
SDP
Catalog No. 5000281613
Encompass_Preferred
Prometon-d14 is the deuterium-labeled analogue of prometon supplied as a stable-isotope reference standard for analytical chemistry. It is intended for use as an internal standard in quantitative mass spectrometry and other analytical methods that require isotopically labeled tracers. CAS number 1219803-45-8; molecular formula C10H5D14N5O.
  • Deuterium-labeled reference for accurate quantitation.
  • Intended for use as an internal standard in LC-MS methods.
  • Enables clear mass differentiation from native analytes.
  • Provided in small, premeasured quantities suited to analytical workflows.
  • Accompanied by molecular data for traceability in experiments.
Catalog No. 50-002-81613 Supplier Medchemexpress LLC Supplier No. HYW414805S1MG
Prometon-d14 is the deuterium-labeled analogue of prometon supplied as a stable-isotope reference standard for analytical chemistry. It is intended for use as an internal standard in quantitative mass spectrometry and other analytical methods that require isotopically labeled tracers. CAS number 1219803-45-8; molecular formula C10H5D14N5O.
  • Deuterium-labeled reference for accurate quantitation.
  • Intended for use as an internal standard in LC-MS methods.
  • Enables clear mass differentiation from native analytes.
  • Provided in small, premeasured quantities suited to analytical workflows.
  • Accompanied by molecular data for traceability in experiments.