Learn More
MSC Diversey Heavy-Duty Floor Cleaner, pH Neutral, Concentrated, 946 mL
Catalog No. 502816442
Encompass
Diversey Prominence Heavy-Duty Floor Cleaner is a concentrated, water-based solution with a pH of 8.4 (diluted pH of approximately 6.85), designed for the effective removal of soil from a diverse range of flooring materials. Its formulation ensures rapid dirt emulsification and dries without leaving film, making it suitable for industrial and commercial floor maintenance.
  • Quickly emulsifies dirt for fast soil removal
  • Dries film-free with no rinsing
  • Neutral pH when diluted (approximately 6.85)
  • Suitable for vinyl, laminate, linoleum, wood, terrazzo, marble, granite, asphalt, porcelain, brick, travertine, ceramic, concrete, rubber, and slate surfaces
  • Water-based and concentrated formula
  • Citrus scent
Catalog No. 50-281-6442 Supplier MSC Supplier No. 03721552
Diversey Prominence Heavy-Duty Floor Cleaner is a concentrated, water-based solution with a pH of 8.4 (diluted pH of approximately 6.85), designed for the effective removal of soil from a diverse range of flooring materials. Its formulation ensures rapid dirt emulsification and dries without leaving film, making it suitable for industrial and commercial floor maintenance.
  • Quickly emulsifies dirt for fast soil removal
  • Dries film-free with no rinsing
  • Neutral pH when diluted (approximately 6.85)
  • Suitable for vinyl, laminate, linoleum, wood, terrazzo, marble, granite, asphalt, porcelain, brick, travertine, ceramic, concrete, rubber, and slate surfaces
  • Water-based and concentrated formula
  • Citrus scent