Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 100 x 4mm

Catalog No. 502395673 Shop All Phenomenex Products
Encompass

Promosil 5m C18 100 PM951004-0

Catalog No. 50-239-5673 Supplier Phenomenex Inc Supplier No. PM9510040
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Promosil 5m C18 100 PM951004-0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.