Learn More
Medchemexpress LLC HY-114158A 100mg Medchemexpress, Pronase E (Activity≥4000 U/mg) CAS:9036-06-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114158A100MG
Encompass_Preferred
Medchemexpress, HY-114158A 100mg Pronase E (Activity≥4000 U/mg) CAS:9036-06-0 Pronase E (Activity≥4000 U/mg) is a mixture of proteolytic enzymes that is obtained from Streptomyces griseus and could digest protein into individual amino acids. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9775