Learn More
Crescent Chemical Co Inc PRONASE E FROM STREPTOMYCES
SDP
Supplier:  Crescent Chemical Co Inc SE33635.02
Encompass_Preferred
Pronase E from Streptomyces griseus min. 6 DMC-U/mg lyophil. For structural analysis of proteins (1, 2). Suitable for preparation of bacteriophage lambda DNA (3). Size - 1G Storage Conditions - +2 °C TO +8 °C Catalog Number - 33635.02 CAS 9036-06-0
Catalog No. 50-248-059