Learn More
Crescent Chemical Co Inc Pronase E from Streptomyces griseus, 9036-06-0, MFCD00132092, 250mg
SDP
Catalog No. 50248058
Encompass_Preferred
Pronase E from Streptomyces griseus, 9036-06-0, MFCD00132092, 250mg, lyophil. For structural analysis of proteins. Suitable for preparation of bacteriophage lambda DNA. Storage Conditions: +2 °C TO +8 °C
Catalog No. 50-248-058 Supplier Crescent Chemical Co Inc Supplier No. SE33635.01
Pronase E from Streptomyces griseus, 9036-06-0, MFCD00132092, 250mg, lyophil. For structural analysis of proteins. Suitable for preparation of bacteriophage lambda DNA. Storage Conditions: +2 °C TO +8 °C