Learn More
Corning™ Pronto!™ Plus Microarray Systems by CoorsTek™ and Promega™
Catalog No. 07200913 Shop All Corning Products
Change view
Click to view available options
Type:
System 5
System 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
07-200-913 System 6
07-200-912 System 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-200-913 Supplier Corning™ Supplier No. 40056
Incorporate all of the components needed to perform a microarray experiment from RNA isolation to post-hybridization washing
Convenient systems combine Promega SV Total RNA Isolation System, Promega ChipShot™ systems for direct-labeling and cleanup of cDNA (fluorescent dyes not included), and Corning Pronto! Microarray Printing and Hybridization Reagents. Deliver reliable and sensitive microarray results when used with DNA microarrays printed on UltraGAPS™ Slides.
•    Optimized for 5μg of total RNA per labeling reaction
•    Detect as little as 3pg target RNA
•    Produce interslide CV less than 10%
•    Utilize easy-to-follow protocols
TRUSTED_SUSTAINABILITY
Specifications
Type System 6