Learn More
Selleck Chemical LLC Propacetamol hydrochloride S4867-1g
SDP
Catalog No. 503110838
Encompass_Preferred
Propacetamol hydrochloride (PRO) is a water-soluble prodrug of paracetamol (PA) which can be parenterally administered as analgesic for the treatment of postoperative pain acute trauma and gastric and/or intestinal disorders
Catalog No. 50-311-0838 Supplier Selleck Chemical LLC Supplier No. S48671G
Propacetamol hydrochloride (PRO) is a water-soluble prodrug of paracetamol (PA) which can be parenterally administered as analgesic for the treatment of postoperative pain acute trauma and gastric and/or intestinal disorders